siRNA Details
| Gene Symbol: 1110058D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004294.1 | - | 2 | 515 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY704530.1 | - | 2 | 509 | Chr. 17 - 64049862 - 64050667 [-] on Build GRCm38 | |

1110058D11Rik
Genomic Map