siRNA Details
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049375.1 | - | 1 | 2008 | ||
AK049600.1 | - | 1 | 1793 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049375.1 | - | 1 | 2008 | ||
AK049600.1 | - | 1 | 1793 |