siRNA Details
| Gene Symbol: Hnrnpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049375.1 | - | 1 | 2078 | ||
| AK049600.1 | - | 1 | 1863 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnpu| Gene Symbol: Hnrnpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049375.1 | - | 1 | 2078 | ||
| AK049600.1 | - | 1 | 1863 | |||