siRNA Details
| Gene Symbol: Hnrnpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049375.1 | - | 1 | 1728 | ||
| AK049600.1 | - | 1 | 1513 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN664869.1 | - | 1 | 77 | Chr. 1 - 178327031 - 178327554 [-] on Build GRCm38 | |

Hnrnpu
Genomic Map