siRNA Details
| Gene Symbol: Rab11fip4os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045898.2 | 2 | 67 | |||
| GenBank mRNA | AK006156.1 | - | 2 | 110 | ||
| AK019731.1 | - | 2 | 69 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11fip4os2| Gene Symbol: Rab11fip4os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045898.2 | 2 | 67 | |||
| GenBank mRNA | AK006156.1 | - | 2 | 110 | ||
| AK019731.1 | - | 2 | 69 | |||