siRNA Details
| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032829.1 | - | 1 | 1763 | ||
| AK087447.1 | - | 1 | 106 | |||
| AK170212.1 | - | 2 | 3912 | |||
| AK173144.1 | - | 14 | 5505 | |||
| BC051539.1 | - | 1 | 985 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssh1| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032829.1 | - | 1 | 1763 | ||
| AK087447.1 | - | 1 | 106 | |||
| AK170212.1 | - | 2 | 3912 | |||
| AK173144.1 | - | 14 | 5505 | |||
| BC051539.1 | - | 1 | 985 | |||