siRNA Details
| Gene Symbol: Gm36823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134652.1 | - | 496 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ049922.1 | - | 3 | 495 | Chr. 3 - 38427708 - 38443632 [-] on Build GRCm38 | |

Gm36823
Genomic Map