siRNA Details
Gene Symbol: 4930544L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019737.1 | - | 1 | 2224 | ||
AK133210.1 | - | 1 | 2249 | |||
AK133230.1 | - | 1 | 2242 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF105706.1 | - | 213 | Chr. - - [-] on Build |
