siRNA Details
| Gene Symbol: 4930544L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019737.1 | - | 1 | 2307 | ||
| AK133210.1 | - | 1 | 2332 | |||
| AK133230.1 | - | 1 | 2325 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF105706.1 | - | 296 | Chr. - - [-] on Build | ||

4930544L04Rik
Genomic Map