siRNA Details
| Gene Symbol: 4930544L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019737.1 | - | 1 | 1867 | ||
| AK133210.1 | - | 1 | 1892 | |||
| AK133230.1 | - | 1 | 1885 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930544L04Rik| Gene Symbol: 4930544L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019737.1 | - | 1 | 1867 | ||
| AK133210.1 | - | 1 | 1892 | |||
| AK133230.1 | - | 1 | 1885 | |||