siRNA Details
Gene Symbol: 4930509J09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040547.1 | 8 | 1134 | |||
GenBank mRNA | AK015737.1 | - | 8 | 1136 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930509J09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040547.1 | 8 | 1134 | |||
GenBank mRNA | AK015737.1 | - | 8 | 1136 |