siRNA Details
| Gene Symbol: 4930509J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040547.1 | 3, 4 | 268 | |||
| GenBank mRNA | AK015737.1 | - | 3, 4 | 270 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715278.1 | - | 3, 4 | 270 | Chr. 3 - 73362027 - 73481651 [-] on Build GRCm38 | |

4930509J09Rik
Genomic Map