siRNA Details
| Gene Symbol: 4930509J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040547.1 | 8 | 1060 | |||
| GenBank mRNA | AK015737.1 | - | 8 | 1062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930509J09Rik| Gene Symbol: 4930509J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040547.1 | 8 | 1060 | |||
| GenBank mRNA | AK015737.1 | - | 8 | 1062 | ||