siRNA Details
| Gene Symbol: 4930509K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040663.1 | 3 | 1654 | |||
| GenBank mRNA | AK015738.1 | - | 3 | 1655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930509K18Rik| Gene Symbol: 4930509K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040663.1 | 3 | 1654 | |||
| GenBank mRNA | AK015738.1 | - | 3 | 1655 | ||