siRNA Details
| Gene Symbol: Gm16364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045600.1 | - | 2 | 1218 | ||
| BB307427.2 | - | 1 | 594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16364| Gene Symbol: Gm16364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045600.1 | - | 2 | 1218 | ||
| BB307427.2 | - | 1 | 594 | |||