siRNA Details
| Gene Symbol: Gm30570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135978.1 | - | 4 | 2211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm30570| Gene Symbol: Gm30570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135978.1 | - | 4 | 2211 | ||