siRNA Details
| Gene Symbol: 5830453J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018006.1 | - | 1 | 123 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717937.1 | - | 1 | 123 | Chr. 11 - 46377713 - 46378581 [-] on Build GRCm38 | |

5830453J16Rik
Genomic Map