siRNA Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034204.1 | - | 2985 | |||
| AK150567.1 | - | 14 | 2507 | |||
| AK151312.1 | - | 14 | 2507 | |||
| AK152811.1 | - | 14 | 2508 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB080497.2 | - | 1 | 209 | Chr. 8 - 41339838 - 41340507 [-] on Build GRCm38 | |

Asah1
Genomic Map