siRNA Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035405.1 | - | 2279 | |||
| AK048320.1 | - | 2358 | ||||
| BC002310.1 | - | 236 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp652| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035405.1 | - | 2279 | |||
| AK048320.1 | - | 2358 | ||||
| BC002310.1 | - | 236 | ||||