siRNA Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035405.1 | - | 1562 | |||
| AK048320.1 | - | 1641 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI735307.1 | - | 1 | 495 | Chr. 11 - 95770568 - 95771470 [+] on Build GRCm38 | |

Zfp652
Genomic Map