siRNA Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035405.1 | - | 2414 | |||
| AK048320.1 | - | 2493 | ||||
| BC002310.1 | - | 371 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD350834.1 | - | 1 | 561 | Chr. 11 - 95771355 - 95772101 [+] on Build GRCm38 | |

Zfp652
Genomic Map