siRNA Details
| Gene Symbol: Gm2464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140446.1 | - | 2 | 761 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ445306.1 | - | 2 | 550 | Chr. 3 - 13470439 - 13471619 [-] on Build GRCm38 | |

Gm2464
Genomic Map