siRNA Details
| Gene Symbol: 4931403E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045306.1 | 4 | 414 | |||
| GenBank mRNA | AK016426.1 | - | 4 | 415 | ||
| BY716074.1 | - | 4 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931403E22Rik| Gene Symbol: 4931403E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045306.1 | 4 | 414 | |||
| GenBank mRNA | AK016426.1 | - | 4 | 415 | ||
| BY716074.1 | - | 4 | 415 | |||