siRNA Details
| Gene Symbol: Gm29154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131742.1 | - | 4 | 331 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706784.1 | - | 4 | 331 | Chr. 9 - 103919501 - 103959421 [-] on Build GRCm38 | |

Gm29154
Genomic Map