siRNA Details
| Gene Symbol: 4933429O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033783.1 | 1 | 13 | |||
| GenBank mRNA | AK006575.1 | - | 1 | 15 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK044555.1 | - | 301 | Chr. - - [-] on Build | ||

4933429O19Rik
Genomic Map