siRNA Details
| Gene Symbol: 4632428C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033631.1 | 3 | 2178 | |||
| GenBank mRNA | AK085759.1 | - | 3 | 2205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4632428C04Rik| Gene Symbol: 4632428C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033631.1 | 3 | 2178 | |||
| GenBank mRNA | AK085759.1 | - | 3 | 2205 | ||