siRNA Details
| Gene Symbol: Gm31463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053231.1 | - | 2 | 230 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB664580.1 | - | 2 | 225 | Chr. 8 - 8576528 - 8578638 [+] on Build GRCm38 | |

Gm31463
Genomic Map