siRNA Details
| Gene Symbol: Miat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003718.2 | 7 | 2620 | |||
| NR_033657.1 | 7 | 2664 | ||||
| GenBank mRNA | AB300594.1 | - | 7 | 2607 | ||
| AK053540.1 | - | 7 | 2608 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Miat| Gene Symbol: Miat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003718.2 | 7 | 2620 | |||
| NR_033657.1 | 7 | 2664 | ||||
| GenBank mRNA | AB300594.1 | - | 7 | 2607 | ||
| AK053540.1 | - | 7 | 2608 | |||