siRNA Details
| Gene Symbol: 0610033M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002748.1 | - | 2 | 62 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703254.1 | - | 2 | 59 | Chr. 6 - 51401060 - 51402217 [-] on Build GRCm38 | |

0610033M10Rik
Genomic Map