siRNA Details
| Gene Symbol: Gm33035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135793.1 | - | 4 | 1084 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm33035| Gene Symbol: Gm33035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135793.1 | - | 4 | 1084 | ||