siRNA Details
| Gene Symbol: 1700037J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006623.1 | - | 1 | 60 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706620.1 | - | 1 | 59 | Chr. 9 - 16599380 - 16599802 [+] on Build GRCm38 | |

1700037J18Rik
Genomic Map