siRNA Details
| Gene Symbol: 1700037N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006624.1 | - | 1 | 1048 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700037N05Rik| Gene Symbol: 1700037N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006624.1 | - | 1 | 1048 | ||