siRNA Details
| Gene Symbol: Gm33056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136014.1 | - | 4 | 365 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ052885.1 | - | 4 | 365 | Chr. 10 - 19290989 - 19325354 [+] on Build GRCm38 | |

Gm33056
Genomic Map