siRNA Details
| Gene Symbol: Gm3942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036450.1 | - | 2 | 280 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB629352.1 | - | 2 | 280 | Chr. 10 - 112837752 - 112845385 [+] on Build GRCm38 | |

Gm3942
Genomic Map