siRNA Details
| Gene Symbol: Gm20187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045052.1 | 3 | 1231 | |||
| GenBank mRNA | AK084401.1 | - | 3 | 1230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20187| Gene Symbol: Gm20187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045052.1 | 3 | 1231 | |||
| GenBank mRNA | AK084401.1 | - | 3 | 1230 | ||