siRNA Details
| Gene Symbol: Gm15866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079714.1 | - | 2, 3 | 138 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB634505.1 | - | 2, 3 | 138 | Chr. 5 - 43557521 - 43601730 [-] on Build GRCm38 | |

Gm15866
Genomic Map