siRNA Details
| Gene Symbol: Gm15866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079714.1 | - | 5 | 879 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB200474.2 | - | 2 | 663 | Chr. 5 - 43557289 - 43599223 [-] on Build GRCm38 | |

Gm15866
Genomic Map