siRNA Details
Gene Symbol: Airn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048015.1 | - | 1 | 189 | ||
AK051797.1 | - | 1 | 637 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB657411.1 | - | 1 | 637 | Chr. 17 - 12763333 - 12763982 [+] on Build GRCm38 |
