siRNA Details
| Gene Symbol: A930005N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020813.1 | - | 1 | 1008 | ||
| BB709254.1 | - | 1 | 317 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930005N03Rik| Gene Symbol: A930005N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020813.1 | - | 1 | 1008 | ||
| BB709254.1 | - | 1 | 317 | |||