siRNA Details
| Gene Symbol: C330026H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037477.1 | - | 1 | 2637 | ||
| AK082813.1 | - | 2 | 300 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB651998.1 | - | 2 | 300 | Chr. 19 - 41005779 - 41006574 [-] on Build GRCm38 | |

C330026H20Rik
Genomic Map