siRNA Details
| Gene Symbol: Gm14133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086556.1 | - | 2 | 547 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB663248.1 | - | 2 | 544 | Chr. 2 - 149754466 - 149798722 [-] on Build GRCm38 | |

Gm14133
Genomic Map