siRNA Details
Gene Symbol: 1600023N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073433.1 | 1 | 905 | |||
GenBank mRNA | AK005529.1 | - | 1 | 888 | ||
AK083048.1 | - | 1 | 906 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1600023N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073433.1 | 1 | 905 | |||
GenBank mRNA | AK005529.1 | - | 1 | 888 | ||
AK083048.1 | - | 1 | 906 |