siRNA Details
| Gene Symbol: 1600023N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073433.1 | 1 | 526 | |||
| GenBank mRNA | AK005529.1 | - | 1 | 509 | ||
| AK083048.1 | - | 1 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1600023N17Rik| Gene Symbol: 1600023N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073433.1 | 1 | 526 | |||
| GenBank mRNA | AK005529.1 | - | 1 | 509 | ||
| AK083048.1 | - | 1 | 527 | |||