siRNA Details
| Gene Symbol: Gm38404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073373.1 | 2 | 303 | |||
| GenBank mRNA | AK051577.1 | - | 2 | 304 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB656042.1 | - | 2 | 304 | Chr. 6 - 127298043 - 127307967 [-] on Build GRCm38 | |

Gm38404
Genomic Map