siRNA Details
| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037324.1 | - | 1 | 1188 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI989011.1 | - | 1 | 260 | Chr. 1 - 64577315 - 64577914 [+] on Build GRCm38 | |

Creb1
Genomic Map