siRNA Details
Gene Symbol: 2210408F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040257.1 | 2 | 235 | |||
GenBank mRNA | BC021953.1 | - | 2 | 142 | ||
BY241377.1 | - | 2 | 236 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2210408F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040257.1 | 2 | 235 | |||
GenBank mRNA | BC021953.1 | - | 2 | 142 | ||
BY241377.1 | - | 2 | 236 |