siRNA Details
| Gene Symbol: 2210408F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040257.1 | 2 | 212 | |||
| GenBank mRNA | BC021953.1 | - | 2 | 119 | ||
| BY241377.1 | - | 2 | 213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210408F21Rik| Gene Symbol: 2210408F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040257.1 | 2 | 212 | |||
| GenBank mRNA | BC021953.1 | - | 2 | 119 | ||
| BY241377.1 | - | 2 | 213 | |||