siRNA Details
| Gene Symbol: Gm3625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK172237.1 | - | 1 | 404 | ||
| AW124459.1 | - | 1 | 502 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK076489.1 | - | 1191 | Chr. - - [-] on Build | ||

Gm3625
Genomic Map