siRNA Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076483.1 | - | 1 | 841 | ||
| BC048860.1 | - | 1 | 2028 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV250368.2 | - | 1 | 50 | Chr. 5 - 121441780 - 121442408 [+] on Build GRCm38 | |

Naa25
Genomic Map