siRNA Details
| Gene Symbol: 3110021N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014069.1 | - | 1 | 1152 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF471482.1 | - | 1 | 300 | Chr. 4 - 108780911 - 108781409 [+] on Build GRCm38 | |

3110021N24Rik
Genomic Map